Sodium bicarbonate : Sodium carbonate concentrate (64 mM : 20 mM) in water IC eluent concentrate (20x) for Metrosep A Supp 5, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
62414-1L 62414-2.5L
62414-1LEA 67.4 EUR
62414-1L 62414-2.5L
Sodium bicarbonate : Sodium carbonate concentrate (64 mM : 20 mM) in water IC eluent concentrate (20x) for Metrosep A Supp 5, Supelco®
Sodium bicarbonate : Sodium carbonate
The “bicarbonate concentrate” is 1-molar solution of sodium bicarbonate (the saturation temperature of the 1-molar solution is 15°C). It is highly advantageous as it is temperature dependent and its subsequent dilution with water can be appropriately controlled
Prepared with sodium carbonate, sodium bicarbonate and high purity water (18.2 MΩ, 0.2 μm filtered)
Certificaten: This eluent concentrate is application tested. It is certified in accordance with ISO Guide 31.