Natriumcarbonaat 72 mM in water eluent concentrate for IC (20x) for Metrosep A Supp 7, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Calcined soda, Carbonic acid disodium salt
72784-1L 72784-2.5L
72784-1LEA 64.2 EUR
72784-1L 72784-2.5L
Natriumcarbonaat 72 mM in water eluent concentrate for IC (20x) for Metrosep A Supp 7, Supelco®
Natriumcarbonaat
Formule: Na₂CO₃
MW: 105.99 g/mol
Kookpunt: 1600
Smeltpunt: 854
MDL Nummer: MFCD00003494
CAS nummer: 497-19-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=855a792cc3b0a5c53bd8d8a8e3cd57193171b41f&vid=c04334e39dee2f7ffda08bd1059a0c1714af8802&prid=web&clid=SAPDAM