VWR® Chem-Resin™, Pipette Tips
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
613-6447EA 46.5 EUR
613-6447 613-6450 613-6472 613-6475 613-6477 613-6478 613-6480 613-6481 613-6484 613-6486
VWR® Chem-Resin™, Pipette Tips
Pipettips
These tips provides a solution for working with challenging strong polar solvents or highly electronegative reagents.
  • More neutral surface tension
  • Manual tips featuring Chem-Resin™
The reduced hydrophobicity on the surface of the tip facilitates low volume draw and drip-free dispense of these classes of chemicals.