TISAB IV Solution for Determination of Fluoride, Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34574-500ML
34574-2.5L
34574-500MLEA
120
EUR
34574-500ML
34574-2.5L
TISAB IV Solution for Determination of Fluoride, Supelco®
Buffers
Buffers voor aanpassing van ionische sterkte
Total ionic strength adjustment buffers (TISAB), is generally used for the microdetermination of fluorides.
TISAB IV was used as a metal ion complexing and ionic concentration adjustment agent, in an experiment investigating the increase in fluoride release, from glass ionomer cement, after coating the discs of glass ionomer with 10% silver fluoride (AgF).