Indicator buffer tablets for the determination of water hardness with Titriplex® solutions, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.08430.0500 1.08430.1000
1.08430.0500EA 37.61 EUR
1.08430.0500 1.08430.1000
Indicator buffer tablets for the determination of water hardness with Titriplex® solutions, Supelco®
Milieutestkits
  • Tablet disintegration time (at 25 °C) ≤ 150 s
  • Tablet weight: 0,20 g ≤ w ≤ 0,23 g