Multi-channel pipettes, mechanical, variable volume, Transferpette® S -8/-12
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
705900 705912 705910 705908 705906 705920 705932 705930 705928 705926 998360
613-6851EA 733 EUR
613-6851 613-6852 613-6853 613-6854 613-6855 613-6856 613-6857 613-6858 613-6859 613-6860 BRND998360
Multi-channel pipettes, mechanical, variable volume, Transferpette® S -8/-12
Pipetten Micropipetten
The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Due to its perfect ergonomics, the Transferpette® S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands.
  • True one-hand operation for right- and left-handed users: Easily set the volume, pipette and eject the tip
  • Colour code on volume adjustment wheel, shaft and volume-change protection
  • Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
  • Easy insertion and ejection of tips due to O-rings on multi-channel pipettes
  • Universal tip cone
  • Fully autoclavable at 121 °C
The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette® S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.