Oversleeves, LDPE
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113-9673EA 86.7 EUR
113-9673 113-9678 113-9680 113-9679 113-9677
Oversleeves, LDPE
Wegwerpkleding
Oversleeves made of 20 or 40 µm LDPE film.
  • Latex-free elastic
  • Length: 400 x 200 mm
Verpakking: 2000 pieces/carton.