Micro-cavity plates, Corning® Elplasia®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4440 4441 4442 4444 4445 4446 4447
CORN4440EA 593.82 EUR
CORN4440 CORN4441 391-1006 CORN4444 CORN4445 CORN4446 CORN4447
Micro-cavity plates, Corning® Elplasia®
Microplaten Celcultuurplaten
Elplasia plates enable researchers to generate high density of spheroids in a scaffold-free model. Use Corning Elplasia plates to generate, culture and analyse your spheroids all in a standard plate footprint.
  • Black opaque side walls reduce well-to-well cross-talk
  • Gamma irradiated
  • Suitable for fluorescent/luminescent assays
  • One year shelf life
These plates are compatible with many cell types and may be used across many applications including: Drug/high throughput screening, cancer/tumour biology, stem cell biology, cell therapy research and 3-D tissue engineering.
Key benefits:
• Create uniform spheroid formation in large volumes with a simple, easy to use 'plug and play' protocol
• Highly reproducible bulk spheroid formation across microcavity wells – from 79 to 15000+ spheroids per well
• Culture a high density of spheroids in one plate under one culture condition
• High density format also generates increased data points, enabling image analysis of multiple spheroids vs. one spheroid per well
• No rinsing required prior to seeding
• Generate and culture spheroids in one plate – for up to 21 or more days
• Increased signal per well without an increase in spheroid size
Elplasia round bottom plates are optimal for bulk spheroid formation, collection and expansion. Round bottom plates are available in 6-, 24- and 96-well formats, and all feature a Corning Ultra-Low Attachment (ULA) surface. Corning ULA surface is proprietary, animal-free, covalently bonded hydrogel surface that is hydrophilic and neutrally charged. The ULA surface promotes the formation and easy harvesting of anchorage-dependent scaffold-free spheroids.
Elplasia square bottom type plates feature a surface with optical qualities suitable for image analysis, making them an ideal solution for clonal selection and high magnification imaging of very small clusters. Square bottom plates are plasma-treated for self-coating and come in 6-, 24-, 96- and 384-well formats.