2-Mercaptoethanol ≥99.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol
97622-10ML 97622-1ML 97622-10X1ML
97622-10MLEA 55.4 EUR
97622-10ML 97622-1ML 97622-10X1ML
2-Mercaptoethanol ≥99.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
2-Mercaptoethanol
Suitable for derivatisation of amine groups.
Formule: HSCH₂CH₂OH
MW: 78.14 g/mol
Kookpunt: 157
Smeltpunt: -100
Vlampunt: 68
MDL Nummer: MFCD00004890
CAS nummer: 60-24-2
UN: 2966
ADR: 6.1 II
Merck Index: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50a0997da05082d8384dbdad2b0f714fa5823da3&vid=940b400110902081cb91993d15b274ca3aea6670&prid=web&clid=SAPDAM