ESD safety shoes, Boa® fit system, uvex 2 trend, 6938
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6938.1/38 6938.1/39 6938.1/40 6938.1/41 6938.1/42 6938.1/43 6938.1/44 6938.1/45 6938.1/46 6938.1/47 6938.1/48 6938.1/49 6938.1/50 6938.1/51 6938.1/52 6938.2/38 6938.2/39 6938.2/40 6938.2/41 6938.2/42 6938.2/43 6938.2/44 6938.2/45 6938.2/46 6938.2/47 6938.2/48 6938.2/49 6938.2/50 6938.2/51 6938.2/52 6938.3/38 6938.3/39 6938.3/40 6938.3/41 6938.3/42 6938.3/43 6938.3/44 6938.3/45 6938.3/46 6938.3/47 6938.3/48 6938.3/49 6938.3/50 6938.3/51 6938.3/52
UVEX6938.1/38EA 81.6 EUR
UVEX6938.1/38 UVEX6938.1/39 UVEX6938.1/40 UVEX6938.1/41 UVEX6938.1/42 UVEX6938.1/43 UVEX6938.1/44 UVEX6938.1/45 UVEX6938.1/46 UVEX6938.1/47 UVEX6938.1/48 UVEX6938.1/49 UVEX6938.1/50 UVEX6938.1/51 UVEX6938.1/52 UVEX6938.2/38 UVEX6938.2/39 UVEX6938.2/40 UVEX6938.2/41 UVEX6938.2/42 UVEX6938.2/43 UVEX6938.2/44 UVEX6938.2/45 UVEX6938.2/46 UVEX6938.2/47 UVEX6938.2/48 UVEX6938.2/49 UVEX6938.2/50 UVEX6938.2/51 UVEX6938.2/52 UVEX6938.3/38 UVEX6938.3/39 UVEX6938.3/40 UVEX6938.3/41 UVEX6938.3/42 UVEX6938.3/43 UVEX6938.3/44 UVEX6938.3/45 UVEX6938.3/46 UVEX6938.3/47 UVEX6938.3/48 UVEX6938.3/49 UVEX6938.3/50 UVEX6938.3/51 UVEX6938.3/52
ESD safety shoes, Boa® fit system, uvex 2 trend, 6938
Schoenen en laarzen
Sporty S1P safety shoe with wide fit.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable, perforated microvelours
  • Reflective details for enhanced visibility
  • Boa® Fit System for individual adjustment and optimum fit all day long
Safety shoes feature a penetration resistant steel midsole and steel toe cap, as well as an ergonomically designed outsole that offers excellent slip resistance. A stabilising uvex anti-twist heel cap is designed to prevent ankle twisting. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/blue
Certificaten: EN 61340 (ESD); EN ISO 20345:2011, S1P, SRC