ESD safety sandals, Velcro® fastening, uvex 2 trend, 6906
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6906.1/38 6906.1/39 6906.1/40 6906.1/41 6906.1/42 6906.1/43 6906.1/44 6906.1/45 6906.1/46 6906.1/47 6906.1/48 6906.1/49 6906.1/50 6906.1/51 6906.1/52 6906.2/38 6906.2/39 6906.2/40 6906.2/41 6906.2/42 6906.2/43 6906.2/44 6906.2/45 6906.2/46 6906.2/47 6906.2/48 6906.2/49 6906.2/50 6906.2/51 6906.2/52 6906.3/38 6906.3/39 6906.3/40 6906.3/41 6906.3/42 6906.3/43 6906.3/44 6906.3/45 6906.3/46 6906.3/47 6906.3/48 6906.3/49 6906.3/50 6906.3/51 6906.3/52
UVEX6906.1/38EA 80.1 EUR
UVEX6906.1/38 UVEX6906.1/39 UVEX6906.1/40 UVEX6906.1/41 UVEX6906.1/42 UVEX6906.1/43 UVEX6906.1/44 UVEX6906.1/45 UVEX6906.1/46 UVEX6906.1/47 UVEX6906.1/48 UVEX6906.1/49 UVEX6906.1/50 UVEX6906.1/51 UVEX6906.1/52 UVEX6906.2/38 UVEX6906.2/39 UVEX6906.2/40 UVEX6906.2/41 UVEX6906.2/42 UVEX6906.2/43 UVEX6906.2/44 UVEX6906.2/45 UVEX6906.2/46 UVEX6906.2/47 UVEX6906.2/48 UVEX6906.2/49 UVEX6906.2/50 UVEX6906.2/51 UVEX6906.2/52 UVEX6906.3/38 UVEX6906.3/39 UVEX6906.3/40 UVEX6906.3/41 UVEX6906.3/42 UVEX6906.3/43 UVEX6906.3/44 UVEX6906.3/45 UVEX6906.3/46 UVEX6906.3/47 UVEX6906.3/48 UVEX6906.3/49 UVEX6906.3/50 UVEX6906.3/51 UVEX6906.3/52
ESD safety sandals, Velcro® fastening, uvex 2 trend, 6906
Schoenen en laarzen
Featuring double, individually adjustable counter-traction hook and loop straps and a breathable microvelour upper. Safety Sandals are suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Penetration resistant steel midsole and steel toe cap
  • Ergonomically designed outsole with excellent slip resistance
  • Stabilising uvex anti-twist heel cap to prevent ankle twisting
  • Reflective details for enhanced visibility
Safety Sandals feature a non-marking PUR outsole and ergonomically designed tread. A two-layer system with abrasion-resistant outsole guarantees excellent walking comfort, as well as excellent slip resistance. Tread is self-cleaning.
Colour: Black/grey/orange
Certificaten: EN 61340 (ESD); EN ISO 20345:2011; S1P, SRC