ESD safety shoes, lace-up, uvex 2 trend, 6934, S3
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6934.3/42 6934.3/43 6934.3/44 6934.3/45 6934.3/46 6934.3/47 6934.3/48 6934.3/49 6934.3/50 6934.3/51 6934.3/52 6934.1/38 6934.1/39 6934.1/40 6934.1/41 6934.1/42 6934.1/43 6934.1/44 6934.1/45 6934.1/46 6934.1/47 6934.1/48 6934.1/49 6934.1/50 6934.1/51 6934.1/52 6934.2/38 6934.2/39 6934.2/40 6934.2/41 6934.2/42 6934.2/43 6934.2/44 6934.2/45 6934.2/46 6934.2/47 6934.2/48 6934.2/49 6934.2/50 6934.2/51 6934.2/52 6934.3/38 6934.3/39 6934.3/40 6934.3/41
UVEX6934.3/42EA 74.7 EUR
UVEX6934.3/42 UVEX6934.3/43 UVEX6934.3/44 UVEX6934.3/45 UVEX6934.3/46 UVEX6934.3/47 UVEX6934.3/48 UVEX6934.3/49 UVEX6934.3/50 UVEX6934.3/51 UVEX6934.3/52 UVEX6934.1/38 UVEX6934.1/39 UVEX6934.1/40 UVEX6934.1/41 UVEX6934.1/42 UVEX6934.1/43 UVEX6934.1/44 UVEX6934.1/45 UVEX6934.1/46 UVEX6934.1/47 UVEX6934.1/48 UVEX6934.1/49 UVEX6934.1/50 UVEX6934.1/51 UVEX6934.1/52 UVEX6934.2/38 UVEX6934.2/39 UVEX6934.2/40 UVEX6934.2/41 UVEX6934.2/42 UVEX6934.2/43 UVEX6934.2/44 UVEX6934.2/45 UVEX6934.2/46 UVEX6934.2/47 UVEX6934.2/48 UVEX6934.2/49 UVEX6934.2/50 UVEX6934.2/51 UVEX6934.2/52 UVEX6934.3/38 UVEX6934.3/39 UVEX6934.3/40 UVEX6934.3/41
ESD safety shoes, lace-up, uvex 2 trend, 6934, S3
Schoenen en laarzen
Sporty S3 safety shoe with wide fit.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Water repellent leather
  • Smooth-running, dirt- and heat-resistant lacing system
  • Foamed polyurethane scuffcap helps increase product life of upper (S3 models)
  • Reflective details for enhanced visibility
Safety Shoes feature a penetration resistant steel midsole (S3 models) and steel toe cap. The ergonomically designed outsole offers excellent slip resistance. Stabilising uvex anti-twist heel cap is designed to prevent ankle twisting. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances
Colour: Black/blue
Certificaten: EN 61340 (ESD); EN ISO 20345:2011, S3, SRC