Goggles, uvex megasonic
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9320.265 9320.281
111-2544EA 28.9 EUR
111-2544 UVEX9320.281
Goggles, uvex megasonic
Ruimzichtbrillen
Megasonic goggle's revolutionary lens design does not restrict the wearer's field of vision, meaning users have the clearest possible view at all times, even under difficult conditions. It has a sporty, ergonomic design and is extremely comfortable to wear, making it a reliable piece of equipment for everyday work.
  • Unique frameless lens design
  • OTG compatible with frame recess for side arms
  • Soft, adaptive face seal for incredible comfort and fit
  • 30 mm wide textile headband for a superior, no pressure fit
  • Easily interchangeable headband and lens
  • Highly compatible with other PPE items
  • Reliable UV 400 protection
  • Equipped with supravision coating technology
The supravision excellence coating provides anti-fog protection on the inside and makes the outside extremely resistant to chemicals and scratches.
Weight: 96 g
Certificaten: EN 166, EN 170 / EN 172