ESD Safety shoes, Boa® Fit System, uvex 2 trend, 6908
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6908.1/38 6908.1/39 6908.1/40 6908.1/41 6908.1/42 6908.1/43 6908.1/44 6908.1/45 6908.1/46 6908.1/47 6908.1/48 6908.1/49 6908.1/50 6908.1/51 6908.1/52 6908.2/38 6908.2/39 6908.2/40 6908.2/41 6908.2/42 6908.2/43 6908.2/44 6908.2/45 6908.2/46 6908.2/47 6908.2/48 6908.2/49 6908.2/50 6908.2/51 6908.2/52 6908.3/38 6908.3/39 6908.3/40 6908.3/41 6908.3/42 6908.3/43 6908.3/44 6908.3/45 6908.3/46 6908.3/47 6908.3/48 6908.3/49 6908.3/50 6908.3/51 6908.3/52
UVEX6908.1/38EA 81 EUR
UVEX6908.1/38 UVEX6908.1/39 UVEX6908.1/40 UVEX6908.1/41 UVEX6908.1/42 UVEX6908.1/43 UVEX6908.1/44 UVEX6908.1/45 UVEX6908.1/46 UVEX6908.1/47 UVEX6908.1/48 UVEX6908.1/49 UVEX6908.1/50 UVEX6908.1/51 UVEX6908.1/52 UVEX6908.2/38 UVEX6908.2/39 UVEX6908.2/40 UVEX6908.2/41 UVEX6908.2/42 UVEX6908.2/43 UVEX6908.2/44 UVEX6908.2/45 UVEX6908.2/46 UVEX6908.2/47 UVEX6908.2/48 UVEX6908.2/49 UVEX6908.2/50 UVEX6908.2/51 UVEX6908.2/52 UVEX6908.3/38 UVEX6908.3/39 UVEX6908.3/40 UVEX6908.3/41 UVEX6908.3/42 UVEX6908.3/43 UVEX6908.3/44 UVEX6908.3/45 UVEX6908.3/46 UVEX6908.3/47 UVEX6908.3/48 UVEX6908.3/49 UVEX6908.3/50 UVEX6908.3/51 UVEX6908.3/52
ESD Safety shoes, Boa® Fit System, uvex 2 trend, 6908
Schoenen en laarzen
ESD Sporty S1P safety shoe for men with wide fit and with slip resistance PU sole.
  • Boa® Fit System for individual adjustment and optimum fit all day long
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable, perforated microvelours
  • Reflective details for enhanced visibility
  • Excellent shock absorption
  • Uvex climazone foot climate management, uvex 1 climate comfort insole, uvex multi-width system helps maintain foot temperature for optimum wearer comfort
Safety Shoes feature a penetration resistant steel midsole and steel toe cap, as well as an ergonomically designed outsole that offers excellent slip resistance. A stabilising uvex anti-twist heel cap is designed to prevent ankle twisting. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
The high-quality outsole materials, sports influenced design and bold colours with maximum comfort and reliable safety. Medium-duty applications. Lining material: distance mesh.
Colour: Black/grey/orange
Certificaten: EN 61340 (ESD); EN ISO 20345:2011, S1P, SRC; certified with uvex medicare.