ESD safety shoes, lace-up, uvex 2 trend, 6937
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6937.7/38 6937.7/39 6937.7/40 6937.7/41 6937.7/42 6937.7/43 6937.7/44 6937.7/45 6937.7/46 6937.7/47 6937.7/48 6937.7/49 6937.7/50 6937.7/51 6937.7/52 6937.8/38 6937.8/39 6937.8/40 6937.8/41 6937.8/42 6937.8/43 6937.8/44 6937.8/45 6937.8/46 6937.8/47 6937.8/48 6937.8/49 6937.8/50 6937.8/51 6937.8/52 6937.9/38 6937.9/39 6937.9/40 6937.9/41 6937.9/42 6937.9/43 6937.9/44 6937.9/45 6937.9/46 6937.9/47 6937.9/48 6937.9/49 6937.9/50 6937.9/51 6937.9/52
UVEX6937.7/38EA 67.6 EUR
UVEX6937.7/38 UVEX6937.7/39 UVEX6937.7/40 UVEX6937.7/41 UVEX6937.7/42 UVEX6937.7/43 UVEX6937.7/44 UVEX6937.7/45 UVEX6937.7/46 UVEX6937.7/47 UVEX6937.7/48 UVEX6937.7/49 UVEX6937.7/50 UVEX6937.7/51 UVEX6937.7/52 UVEX6937.8/38 UVEX6937.8/39 UVEX6937.8/40 UVEX6937.8/41 UVEX6937.8/42 UVEX6937.8/43 UVEX6937.8/44 UVEX6937.8/45 UVEX6937.8/46 UVEX6937.8/47 UVEX6937.8/48 UVEX6937.8/49 UVEX6937.8/50 UVEX6937.8/51 UVEX6937.8/52 UVEX6937.9/38 UVEX6937.9/39 UVEX6937.9/40 UVEX6937.9/41 UVEX6937.9/42 UVEX6937.9/43 UVEX6937.9/44 UVEX6937.9/45 UVEX6937.9/46 UVEX6937.9/47 UVEX6937.9/48 UVEX6937.9/49 UVEX6937.9/50 UVEX6937.9/51 UVEX6937.9/52
ESD safety shoes, lace-up, uvex 2 trend, 6937
Schoenen en laarzen
Sporty S1 safety shoe with wide fit, made from microvelour upper and a dual-density PU sole.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable, perforated microvelours
  • Steel toe cap
  • Reflective details for enhanced visibility
  • Excellent shock absorption
  • High levels of wearer comfort
Safety shoes feature an ergonomically designed outsole with excellent slip resistance, as well as a stabilising uvex anti-twist heel cap to prevent ankle twisting. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/blue
Certificaten: EN 61340 (ESD); EN ISO 20345:2011 S1 SRC