ESD cccupational shoes, lace-up, uvex 1 sport, 6595
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6595.7/35 6595.7/36 6595.7/37 6595.7/40 6595.7/41 6595.7/42 6595.7/43 6595.7/45 6595.7/47 6595.7/48 6595.7/49 6595.7/50 6595.7/51 6595.7/52 6595.8/41
UVEX6595.7/35EA 0 EUR
UVEX6595.7/35 UVEX6595.7/36 UVEX6595.7/37 UVEX6595.7/40 UVEX6595.7/41 UVEX6595.7/42 UVEX6595.7/43 UVEX6595.7/45 UVEX6595.7/47 UVEX6595.7/48 UVEX6595.7/49 UVEX6595.7/50 UVEX6595.7/51 UVEX6595.7/52 UVEX6595.8/41
ESD cccupational shoes, lace-up, uvex 1 sport, 6595
Schoenen en laarzen
Modern, ultra-lightweight, and flexible O1 occupational shoe without toecap.
  • Breathable textile upper material
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Climazone technology for optimum comfort
  • Virtually seam-free construction to eliminate pressure points
  • Ergonomically designed dual density polyurethane outsole for excellent slip resistant
Occupational Shoes offer outstanding wearer comfort due to newly developed climate-optimised, breathable materials. Shoes feature a removeable anti-static comfortable insole, with a moisture transport system and additional shock absorption around the heel and forefoot. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances.
Colour: Black/white
Certificaten: EN 61340 (ESD); EN ISO 20347:2012 O1 FO SRC