uvex i-5 Safety Spectacles
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9183.281 9183.265 9183064
111-2448EA 26.2 EUR
111-2448 111-2449 UVEX9183064
uvex i-5 Safety Spectacles
Brillen
Reliable protection thanks to an innovative and modern design. With an x-tended eyeshield and sideshield, the uvex i-5 can be relied on to protect the eyes in any situation. Sophisticated features ensure superior wearer comfort, even during long periods of use.
  • Soft, flexible nose pad for maximum comfort and a non-slip fit
  • x-tended eyeshield provides additional protection from flying particles, dust, and dirt
  • x-tended sideshield for increased side protection
  • 3-position side arm inclination adapts easily to any facial shape
  • Reliable UV 400 protection
  • Equipped with supravision coating technology
Soft ear pieces (uvex duo Component technology) prevent pressure points and ensure a non-slip fit. The uvex supravision excellence coating provides anti-fog protection on the inside and makes the outside extremely resistant to chemicals and scratches. Soft, extendable x-twist side arm technology offers adaptable, pressure-free comfort and a stable, non-slip fit.