ESD safety sandals, Velcro® fastening, uvex 2 trend, S1P, 6936
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6936.1/38 6936.1/39 6936.1/40 6936.1/41 6936.1/42 6936.1/43 6936.1/44 6936.1/45 6936.1/46 6936.1/47 6936.1/48 6936.1/49 6936.1/50 6936.1/51 6936.1/52 6936.2/38 6936.2/39 6936.2/40 6936.2/41 6936.2/42 6936.2/43 6936.2/44 6936.2/45 6936.2/46 6936.2/47 6936.2/48 6936.2/49 6936.2/50 6936.2/51 6936.2/52 6936.3/38 6936.3/39 6936.3/40 6936.3/41 6936.3/42 6936.3/43 6936.3/44 6936.3/45 6936.3/46 6936.3/47 6936.3/48 6936.3/49 6936.3/50 6936.3/51 6936.3/52
UVEX6936.1/38EA 69.5 EUR
UVEX6936.1/38 UVEX6936.1/39 UVEX6936.1/40 UVEX6936.1/41 UVEX6936.1/42 UVEX6936.1/43 UVEX6936.1/44 UVEX6936.1/45 UVEX6936.1/46 UVEX6936.1/47 UVEX6936.1/48 UVEX6936.1/49 UVEX6936.1/50 UVEX6936.1/51 UVEX6936.1/52 UVEX6936.2/38 UVEX6936.2/39 UVEX6936.2/40 UVEX6936.2/41 UVEX6936.2/42 UVEX6936.2/43 UVEX6936.2/44 UVEX6936.2/45 UVEX6936.2/46 UVEX6936.2/47 UVEX6936.2/48 UVEX6936.2/49 UVEX6936.2/50 UVEX6936.2/51 UVEX6936.2/52 UVEX6936.3/38 UVEX6936.3/39 UVEX6936.3/40 UVEX6936.3/41 UVEX6936.3/42 UVEX6936.3/43 UVEX6936.3/44 UVEX6936.3/45 UVEX6936.3/46 UVEX6936.3/47 UVEX6936.3/48 UVEX6936.3/49 UVEX6936.3/50 UVEX6936.3/51 UVEX6936.3/52
ESD safety sandals, Velcro® fastening, uvex 2 trend, S1P, 6936
Schoenen en laarzen
ESD sporty S1P safety sandals with wide fit, made from microvelour upper and a dual-density PU sole.
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Breathable microvelour upper
  • Double, individually adjustable velcro fastening
  • Reflective details for enhanced visibility
  • Excellent shock absorption
  • High levels of wearer comfort
Safety sandals feature an ergonomically designed outsole that offers excellent slip resistance, as well as a stabilising uvex anti-twist heel cap designed to prevent ankle twisting. Suitable for people allergic to chrome as they are made from synthetic materials. All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances. Removable, anti-static, comfortable insole, with a moisture transport system and additional shock absorption around the heel and forefoot.
Colour: Blue/black
Certificaten: EN 61340 (ESD); EN ISO 20345:2011; S1P, SRC