Industrial gloves, MIAMI BLUE, TRIUS®
Leverancier: TRIUS SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MIAMIBLUE/7 MIAMIBLUE/8 MIAMIBLUE/11 MIAMIBLUE/12 MIAMIBLUE/10 MIAMIBLUE/9
TRIUMIAMIBLUE/7EA 81.8 EUR
TRIUMIAMIBLUE/7 TRIUMIAMIBLUE/8 TRIUMIAMIBLUE/11 TRIUMIAMIBLUE/12 TRIUMIAMIBLUE/10 TRIUMIAMIBLUE/9
Industrial gloves, MIAMI BLUE, TRIUS®
Handschoenen
These handling gloves feature a natural tawed leather palm and fingertips, blue jersey back, and Velcro® tightening at wrist.
  • High-quality goat leather
  • Breathable back
  • Velcro® tigthtening at wrist
Goat leather handling gloves with breathable cotton back offer high dexterity in dry environments.
Certificaten: EN 388:2016 (2122X), EN 420:2003 + A1:2009 dexterity 5, REACh compliant.
Verpakking: Bundle of 10 pairs with UIS.