Industrial gloves, NEW STAR, TRIUS®
Leverancier: TRIUS SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NEWSTAR/10 NEWSTAR/11 NEWSTAR/7 NEWSTAR/8 NEWSTAR/9
112-6024EA 101 EUR
112-6024 112-6025 112-6027 112-6028 112-6029
Industrial gloves, NEW STAR, TRIUS®
Handschoenen
These full grain cow leather handling gloves are designed for use in dry environments.
  • High quality leather
  • Extra grip
  • Good dexterity
  • Elastic on back
Handling gloves feature all natural cowhide grain leather, with an elasticated wrist band.
Certificaten: EN 388:2016 (3132X), EN 420:2003 + A1:2009 dexterity 5, REACh compliant.
Verpakking: Bundle of 10 pairs with UIS.