Cut resistant gloves, ROCK CUT, TRIUS®
Leverancier: TRIUS SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROCKCUT/8 ROCKCUT/6 ROCKCUT/7 ROCKCUT/10 ROCKCUT/11 ROCKCUT/9
TRIUROCKCUT/8EA 47.9 EUR
TRIUROCKCUT/8 TRIUROCKCUT/6 TRIUROCKCUT/7 TRIUROCKCUT/10 TRIUROCKCUT/11 TRIUROCKCUT/9
Cut resistant gloves, ROCK CUT, TRIUS®
Handschoenen
These cut resistant, economic gloves feature a class C cut-resistant fiber, a salt and pepper finish, and black PU palm coating.
  • High cut resistance
  • Good dexterity
  • Abrasion resistant PU coating
The multi-purpose gloves are designed for routine handling in dry environments.
Certificaten: EN 388:2016 (4443C), EN 420:2003 + A1:2009 dexterity 5, REACh compliant.
Verpakking: Bundle of 10 pairs with UIS.
Opgelet: Not for use in case of heat.