Leather gloves, VANCOUVER, TRIUS®
Leverancier: TRIUS SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VANCOUVER/10 VANCOUVER/11 VANCOUVER/8 VANCOUVER/9
TRIUVANCOUVER/10EA 92.2 EUR
TRIUVANCOUVER/10 TRIUVANCOUVER/11 TRIUVANCOUVER/8 TRIUVANCOUVER/9
Leather gloves, VANCOUVER, TRIUS®
Handschoenen
These extra-warm leather gloves provide comfort in cold and wet weather conditions. They feature a water­repellent cowhide split leather palm, a blue water­repellent cowhide grain leather back, thick acrylic fleece lining, and an elasticated wrist.
  • High grade of leather
  • Ultra-warm
  • Water repellent
  • For cold and wet environments
Vancouver handling gloves are designed for industrial applications in cold environments.
Certificaten: EN 388:2016 (4132X), EN 511:2006 (12X), EN 420:2003 + A1:2009 dexterity 5, REACh compliant.
Verpakking: Bundle of 5 pairs with UIS.