Tube top vacuum filtration systems, Sartolab® RF 50
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
180E01-2 180F01-2 180E01-E8 180F01-E8
514-1194EA 121 EUR
514-1194 514-1195 514-1459 514-1460
Tube top vacuum filtration systems, Sartolab® RF 50
Filters Flesfilters
Sartolab® RF 50 vacuum filtration units are single-use units for the filtration of sample volumes up to 50 ml.
They are available with either a 0,22 or 0,45 µm polyethersulfone membrane.
  • Highest flow rates with an asymmetric membrane and a large filtration surface
  • No loss of protein with a low protein affinity membrane
  • Low dead volume due to an optimized membrane support
  • Minimised risk of contamination with a complete ready-to-use unit (no additional transfer of liquid necessary for storage)
The design of the yellow adapter connecting the funnel to the conical tube ensures a vacuum-tight seal and enables the filtration unit to be used on the Sartolab® MultiStation for filtration of up to 6 samples in parallel with one vacuum source. However, Sartolab® RF 50 can also be used alone by directly connecting the unit’s tubing connector to your vacuum source.
Both membranes provide fast flow rates and ensure low protein binding and extractables. Both versions are individually-packed, sterile, and ready to use. The 0,22 µm version is ideal for sterile filtration of cell cultures, buffers and aqueous solutions, while the 0,45 µm version is ideal for clarification.
Pressure: -350...-700 mbar
Leveringsinformatie: The Sartolab® RF 50 vacuum filtration unit comprises a funnel with dust cover, a 50 ml conical tube with graduation and writing field, a tubing connector for vacuum connection and a screw cap for storage of your filtrate (individually wrapped to maintain sterility).