1-Butyl-2-pyrrolidone, EMPLURA®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: N-Butyl-2-azacyclopentanone, 1-Butylpyrrolidin-2-one
1.03818.1000 1.03818.2500 1.03818.6025
1.03818.1000EA 61.9 EUR
1.03818.1000 1.03818.2500 1.03818.6025
1-Butyl-2-pyrrolidone, EMPLURA®
1-Butyl-2-pyrrolidone
1-Butylpyrrolidin-2-one is a safer alternative to N-Methyl-2-pyrrolidone (NMP), N,N-Dimethylacetamide (DMA), Dimethyl sulfoxide (DMSO) and N,N-Dimethylformamide (DMF), which face increasing regulatory pressure. As opposed to NMP, DMF and DMA, 1-Butylpyrrolidin-2-one is not classified as developmentally reprotoxic.
  • Excellent solvency power and water miscibility
  • High boiling point
  • High chemical and thermal stability
  • Not classified as a developmentalor geno-toxin
  • Inherently bio-degradable
  • Lower volatility compared to NMP
  • Reliable alternative for REACH-restricted DMA, DMF, NMP
Formule: C₈H₁₅NO
MW: 141.21 g/mol
MDL Nummer: MFCD00020876
CAS nummer: 3470-98-2