Pipette holders for Transferpette® S
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
704807 704812 704811
BRND704807EA 62.2 EUR
BRND704807 BRND704812 BRND704811
Pipette holders for Transferpette® S
Pipethouders
Keep your Transferpette® S pipettes close at hand with these storage options.
  • Designed to fit 2020 and later Transferpette® S pipettes (serial numbers 20A and higher)
The carousel rack holds up to six pipettes. The hooks can be replaced to fit older HandyStep® touch or HandyStep® S instruments.
The shelf mount allows easy mounting of one Transferpette® S single- or multichannel instrument to a shelf or bench edge. It can also be used to retrofit older BRAND® Transferpette® S carousels to newer instruments.
The wall mount allows easy mounting ofMore Product Information