Glove liners, S-BCRL series, sterile cut resistant, BioClean
Leverancier: Ansell-Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
112-5741EA 1510 EUR
112-5741 112-5742 112-5743 112-5744 112-5745
Glove liners, S-BCRL series, sterile cut resistant, BioClean
Handschoenen
BioClean sterile cut-resistant glove liners feature Dyneema® Diamond yarn and provide cut resistance and protection during rigorous procedures.
  • Level II cut resistance, silicone-free
  • Optimal dexterity, comfort and fit
  • Recommended to be worn between two cleanroom gloves
  • Can be autoclaved for up to five cycles at 121 °C for 30 minutes
  • Made from ultra-high molecular weight polyethylene (UHMWPE)
Specifically constructed for optimal dexterity, comfort and fit, these glove liners also offer ANSI cut level II protection. These cut-resistant glove liners are designed to be worn between two cleanroom gloves to offer cut protection when handling sharp objects or cleaning apparatus which pose a cut risk.
Certificaten: Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications. Conforms to Category 2 - Personal Protective Equipment Regulation (EU) 2016/425. Processed in an NEBB certified ISO Class 4 environment. Complies with the requirements of Glove Standard EN 420:2003 + A1:2009
Sterilised in accordance with ISO 11137-1:2006.
Verpakking: Glove liners are packed 1 pair per inner PE wallet; 1 wallet per sealed EasyTear PE pouch; 10 pouches per sealed outer PE bag; 10 outer bags per lined carton (100 pairs).
Opgelet: No liner provides complete protection against cuts. Use care when handling sharp objects. No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and the environment in which they will be used.