VWR®, Flash point reference standard, Cleveland Open Cup
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99161.001EA 247 EUR
99161.001 99162.001 99163.001 99164.001 99165.001
VWR®, Flash point reference standard, Cleveland Open Cup
Standaarden Standaarden voor de petroleumindustrie
COC flash point standards are tested and certified in accordance with ASTM D92 'Standard Test Method for Flash and Fire Points by Cleveland Open Cup Tester.'
  • Certified according to ISO 17025/ISO 17034 under UKAS accreditation
  • One bottle per test ensures zero contamination/high-end volatile loss
  • Fully traceable to international standards
  • Supplied with MSDS
  • Tamper-evident security packaging
  • 12 month shelf life
These UKAS certified standards are specifically designed to be used for regular instrument performance checks. Packed uniquely in 3×80 ml 'single shot' bottles ensures the highest level of sample integrity. An economical, time saving addition to any laboratory concerned with the generation of valid analytical test data. Typically used in, but not limited to, the analysis of petroleum products with flash points above 79 °C (175 °F) and below 400 °C (752 °F).
Certificaten: UKAS ISO 17025 and ISO 17034.
Opgelet: The flash point given in the ordering table below is the nominal flash point - actual batch values are given in the certification documents supplied with the product and also on the label.
Order Now
Specificaties
Environmentally Preferable