Desiccator lids, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
244305702 244306604
467-1475EA 128 EUR
467-1475 467-1485
Desiccator lids, DURAN®
Deksels Exsiccatordeksels
DURAN®, clear borosilicate glass, 3.3. Suitable for WERTEX type vacuum desiccators.
  • With special tube (NS 24/29, Type WERTEX)
  • With ring-grooved flange