PTFE coated silicone seals, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
292489102 292481456 292482958
SCOT292489102EA 92.2 EUR
SCOT292489102 SCOT292481456 SCOT292482958
PTFE coated silicone seals, DURAN®
Tussenschoten
High-quality silicone seals developed for the needs of the pharmaceutical and biotech industry.
  • Cleanroom packaged, supplied with Certificate of Analysis (CoA), USP <88> Class VI Pharmaceutical grade materials
  • Recommended usable temperature range is –196 to 200 °C
  • Sterilisable via steam/autoclave, chemical, or dry heat
  • Lab washer compatible and microwave safe
High-quality, PTFE coated silicone seals are designed for use with DURAN® PURE Premium Screw Caps. The silicone portion is light ivory in color, while the PTFE portion is gray.
Verpakking: 10 pieces supplied per bag.