VWR®, Racks for Test Tubes
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
212-0713EA 35.5 EUR
212-0713 212-0714 212-0715 212-0716 212-0717 212-0718 212-0719 212-0720 212-0721 212-0722 212-0723 212-0724 212-0725
VWR®, Racks for Test Tubes
Rekken Tube Racks
These test tube racks are either made from rust-proof, electrolytically polished stainless steel or from polyamide coated steel.
  • Very good chemical and temperature resistance
  • Polyamide coated racks are heat resistant up to 80 °C
  • Robust and stable, easy to clean
  • Hole size 18×18 mm