Reagent reservoirs
Leverancier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
120753 120750 120752
HEAT120753EA 88.8 EUR
HEAT120753 HEAT120750 HEAT120752
Reagent reservoirs
Reagensreservoirs
These disposable reagent reservoirs have a fresh and modern design with enhanced features to streamline pipetting procedures.
  • Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sterile, sealed in bags and electron beam sterilised
  • DNase, RNase and pyrogen free
Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Certificaten: Quality certificate available on request.
Verpakking: Sealed in easy-to-open bags. No scissors needed to open.