Tetrahydrofuraan, anhydrous ≥99.9% unstabilised for applications requiring low water content, over molecular sieves <0.0025% H₂O, Riedel-de Haën™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Butylene oxide, THF, Diethylene oxide
Packed in septum-sealed bottle.
66200-1L 66200-2.5L
HONC66200-1LEA 274 EUR
HONC66200-1L HONC66200-2.5L
Tetrahydrofuraan, anhydrous ≥99.9% unstabilised for applications requiring low water content, over molecular sieves <0.0025% H₂O, Riedel-de Haën™
Tetrahydrofuraan
Formule: C₄H₈O
Kookpunt: 65 °C (1,013 hPa)
Smeltpunt: –108 °C
Dichtheid: 0,89 g/cm3 (20 °C)
Vlampunt: –21 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005356
CAS nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (GC) Min. 99.9 %
Water (Karl Fischer) Max. 0.0025 %