Pyridine, anhydrous ≥99.8% for applications requiring low water content, over molecular sieves <0.0050% H₂O, Riedel-de Haën™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Azabenzene, Azine
Packed in septum-sealed bottle.
66213-1L 66213-2.5L
HONC66213-1LEA 334 EUR
HONC66213-1L HONC66213-2.5L
Pyridine, anhydrous ≥99.8% for applications requiring low water content, over molecular sieves <0.0050% H₂O, Riedel-de Haën™
Pyridine
Formule: C₅H₅N
MW: 79.1 g/mol
Kookpunt: 115 °C (1.013 hPa)
Smeltpunt: –42 °C
Dichtheid: 0.98 g/cm3 (20 °C)
Vlampunt: 20 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011732
CAS nummer: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (GC) Min. 99.8 %
Water (Karl Fischer) Max. 0.0050 %