Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
37863-1L 37863-6X1L
37863-1LEA 91.1 EUR
37863-1L 37863-6X1L
Chloroform, Hydranal™ solubilizer for Karl Fischer titration, Fluka™
Chloroform
Formule: CHCl₃
MW: 119.38 g/mol
Kookpunt: 60…62 °C (1.013 hPa)
Smeltpunt: –63 °C
Dichtheid: 1.48 g/cm³ (20 °C)
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD00000826
CAS nummer: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 3 III
Bereik: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Suitability (for KF-titration) test passed
Assay of water Max. 0.01 %
Number Of Samples Min. 2