Bidirectionele universele filamenttape 8959, Scotch®
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8959-50MM 8959-75MM
MMMA8959-50MMEA 18.8 EUR
MMMA8959-50MM MMMA8959-75MM
Bidirectionele universele filamenttape 8959, Scotch®
Tape
This is a tough box-sealing tape with exceptional adhesion that also resists nicks, abrasions, moisture, scuffing, and center seam splitting in both longitudinal and horizontal directions.
  • Filaments down the length and across the width for extra security; tape is difficult to hand-tear in any direction
  • Fiberglass filaments run in bi-directional pattern, providing high tensile strength across the width and length of the tape
  • Resists nicks, abrasions, moisture, and scuffing
  • Aggressive adhesive sticks to fiberboard and other substrates
  • Transparency provides