Washi afplaktape 244, Scotch®
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
244-18MM 244-24MM 244-36MM 244-48MM
819-0118EA 11 EUR
819-0118 819-0119 819-0120 819-0121
Washi afplaktape 244, Scotch®
Tape
Deze is voorzien van een gele gladde papieren rug en een 0,085 mm dikke synthetische lijm. Deze tape is bijzonder goed bestand tegen UV en is waterdicht.
  • Excellent UV resistance for outdoor use (max. six weeks)
  • Clean removal, even after six weeks from most surfaces
  • Ultra-flat paint edges and extremely thin backing material
  • Resists paint bleed
  • Easily removed after heating, without leaving residues
  • Temperature-resistant up to 100 °C for 30 minutes
This tape can be used for a wide range of masking applications, especially for where UV resistance andMore Product Information