Signs, COVID-19 VIRUS social distancing and safety signage - keep safe distance
Leverancier: ERGOMAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DSVSIGN10SDAEN DSVSIGN10SDADE DSVSIGN10SDAFR DSVSIGN10SDAES DSVSIGN10SDANL DSVSIGN10SDAPL DSVSIGN15SDAEN DSVSIGN15SDADE DSVSIGN15SDAFR DSVSIGN15SDAES DSVSIGN15SDANL DSVSIGN15SDAPL DSVSIGN30SDAEN DSVSIGN30SDADE DSVSIGN30SDAFR DSVSIGN30SDAES DSVSIGN30SDANL DSVSIGN30SDAPL
ERGODSVSIGN10SDAENEA 241 EUR
ERGODSVSIGN10SDAEN ERGODSVSIGN10SDADE ERGODSVSIGN10SDAFR ERGODSVSIGN10SDAES ERGODSVSIGN10SDANL ERGODSVSIGN10SDAPL ERGODSVSIGN15SDAEN ERGODSVSIGN15SDADE ERGODSVSIGN15SDAFR ERGODSVSIGN15SDAES ERGODSVSIGN15SDANL ERGODSVSIGN15SDAPL ERGODSVSIGN30SDAEN ERGODSVSIGN30SDADE ERGODSVSIGN30SDAFR ERGODSVSIGN30SDAES ERGODSVSIGN30SDANL ERGODSVSIGN30SDAPL
Signs, COVID-19 VIRUS social distancing and safety signage - keep safe distance
Pictogrammen
In this very complicated period of operation, we have organized an efficient and quick solution for floor and/or wall marking signage that is in line with the distance rules and recommendations to be observed.
  • Multiple language options
  • 10cm, 15cm, and 30,5 cm diameter sizes
These signs can be utilized in (but not limited to) hospitals, mass distribution, pharmacies, and any other field that continues to operate during this uncertain period.