Signs, COVID-19 VIRUS social distancing and safety signage - face mask required
Leverancier: ERGOMAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DSVSIGN10SDBEN DSVSIGN10SDBDE DSVSIGN10SDBFR DSVSIGN10SDBES DSVSIGN10SDBNL DSVSIGN10SDBPL DSVSIGN15SDBEN DSVSIGN15SDBDE DSVSIGN15SDBFR DSVSIGN15SDBES DSVSIGN15SDBNL DSVSIGN15SDBPL DSVSIGN30SDBEN DSVSIGN30SDBDE DSVSIGN30SDBFR DSVSIGN30SDBES DSVSIGN30SDBNL DSVSIGN30SDBPL
ERGODSVSIGN10SDBENEA 241 EUR
ERGODSVSIGN10SDBEN ERGODSVSIGN10SDBDE ERGODSVSIGN10SDBFR ERGODSVSIGN10SDBES ERGODSVSIGN10SDBNL ERGODSVSIGN10SDBPL ERGODSVSIGN15SDBEN ERGODSVSIGN15SDBDE ERGODSVSIGN15SDBFR ERGODSVSIGN15SDBES ERGODSVSIGN15SDBNL ERGODSVSIGN15SDBPL ERGODSVSIGN30SDBEN ERGODSVSIGN30SDBDE ERGODSVSIGN30SDBFR ERGODSVSIGN30SDBES ERGODSVSIGN30SDBNL ERGODSVSIGN30SDBPL
Signs, COVID-19 VIRUS social distancing and safety signage - face mask required
Pictogrammen
In this very complicated period of operation, we have organized an efficient and quick solution for floor and/or wall marking signage that is in line with the distance rules and recommendations to be observed.
  • Multiple language options
  • 10cm, 15cm, and 30,5 cm diameter sizes
These signs can be utilized in (but not limited to) hospitals, mass distribution, pharmacies, and any other field that continues to operate during this uncertain period.