Signs, COVID-19 VIRUS social distancing and safety signage - gloves required
Leverancier: ERGOMAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DSVSIGN10SDCEN DSVSIGN10SDCDE DSVSIGN10SDCFR DSVSIGN10SDCES DSVSIGN10SDCNL DSVSIGN10SDCPL DSVSIGN15SDCEN DSVSIGN15SDCDE DSVSIGN15SDCFR DSVSIGN15SDCES DSVSIGN15SDCNL DSVSIGN15SDCPL DSVSIGN30SDCEN DSVSIGN30SDCDE DSVSIGN30SDCFR DSVSIGN30SDCES DSVSIGN30SDCNL DSVSIGN30SDCPL
ERGODSVSIGN10SDCENEA 241 EUR
ERGODSVSIGN10SDCEN ERGODSVSIGN10SDCDE ERGODSVSIGN10SDCFR ERGODSVSIGN10SDCES ERGODSVSIGN10SDCNL ERGODSVSIGN10SDCPL ERGODSVSIGN15SDCEN ERGODSVSIGN15SDCDE ERGODSVSIGN15SDCFR ERGODSVSIGN15SDCES ERGODSVSIGN15SDCNL ERGODSVSIGN15SDCPL ERGODSVSIGN30SDCEN ERGODSVSIGN30SDCDE ERGODSVSIGN30SDCFR ERGODSVSIGN30SDCES ERGODSVSIGN30SDCNL ERGODSVSIGN30SDCPL
Signs, COVID-19 VIRUS social distancing and safety signage - gloves required
Pictogrammen
In this very complicated period of operation, we have organized an efficient and quick solution for floor and/or wall marking signage that is in line with the distance rules and recommendations to be observed.
  • Multiple language options
  • 10cm, 15cm, and 30,5 cm diameter sizes
These signs can be utilized in (but not limited to) hospitals, mass distribution, pharmacies, and any other field that continues to operate during this uncertain period.