First aid burn kit, Cederroth
Leverancier: Orkla
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
118-0499EA 211 EUR
118-0499
First aid burn kit, Cederroth
Eerste-hulpbenodigdheden Brandwondenverzorging
A mobile first aid kit with products for handling burns. Burn gel products provide cooling and pain relief for first- and second-degree burns. Burn gel dressings come in three sizes, including a large dressing/face mask.
  • Easy to use with clear instructions
  • Practical handle
  • Marked with flame and orange zipper for increased visibility
  • Transparent foldable pockets gives a clear overview of the contents
Suitable for workplaces with high risk of burns, such as environments featuring open fire, flames, hot surfaces, steam, hot fluids and ovens. Typical workplaces include building, paper, metal, chemical and HoReCa industries.
Contents:
1× Burn gel, 100 ml
1× Burn gel dressing/face mask, 30×40 cm
2× Burn gel dressing, 20×20 cm
4× Burn gel dressing, 10×10 cm
1× Cederroth 4-in-1 Bloodstopper
1× Fixing net
6× Elastic fixing bandage
2 pairs of gloves
1× Scissors
Contents list
First aid and burn instructions