Industrial gloves, phynomic airLite A ESD
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6003806 6003807 6003808 6003809 6003810 6003811 6003812
UVEX6003806EA 56.8 EUR
UVEX6003806 UVEX6003807 UVEX6003808 UVEX6003809 UVEX6003810 UVEX6003811 UVEX6003812
Industrial gloves, phynomic airLite A ESD
Handschoenen
The most lightweight safety glove within its range with ESD-Function according to DIN 16350.
  • An excellent combination of high dexterity, touch sensitivity, lightweight design, and breathability
  • Aqua-polymer foam airLite coating on palm and fingertips
  • Outer material made from polyamide, elastane, and carbon
  • Use on almost all screens, tablets, and mobile phones
  • Excellent grip in dry and slightly damp areas
The new Aqua-Polymer foam airLite coating, together with a very thin liner (18 gauge), provides very good dexterity for precision work and handling of very small pieces, in dry and slightly damp areas. The additional touchscreen ability make the phynomic airLite A suitable on all touchscreens, tablets and mobile phones. These safety glove are free of harmful substances, with a skin compatibility that is dermatological approved (proDerm). The gloves are therefore also very useful for allergic persons.
Certificaten: EN 388 (3110X), DIN EN 16350:2014
Verpakking: 10 pairs per bundle.