Mouse Recombinant EGF (from E. coli)
Leverancier: Shenandoah Biotechnology
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
200-53AF-100UG 200-53AF-500UG 200-53AF-5UG
SHBT200-53AF-100UGEA 78.1 EUR
SHBT200-53AF-100UG SHBT200-53AF-500UG SHBT200-53AF-5UG
Mouse Recombinant EGF (from E. coli)
Eiwitten en peptiden
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
  • High quality
  • Low endotoxin
  • Affordable
Opgelet: This product is for research use only and is not for use in humans.