Dry swab with transport tube, Q-SWAB™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T307-10A T307-5A
SIMPT307-10AEA 60.2 EUR
SIMPT307-10A SIMPT307-5A
Dry swab with transport tube, Q-SWAB™
Wattenstaafjes Wattenstaafjes in buisjes
Sterile specimen collection swab.
  • Polypropylene tubes are translucent
  • Cap equipped with silicone washer
  • Non graduated
Q-Swab™ is self-standing, but features a round bottom. The polyester swab holds securely inside the screw cap, making it ideal for specimen collection and protection from contamination. The polypropylene tubes are translucent, making it easy to see through. The cap features an exclusive silicone washer fitted inside to ensure a positive seal at any temperature.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904