ESD safety ankle boots, lace-up, L10, Vintage Pirate
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
762071/47 762071/37 762071/38 762071/39 762071/40 762071/41 762071/42 762071/43 762071/44 762071/45 762071/46
114-8842EA 93.9 EUR
114-8842 114-8832 114-8833 114-8834 114-8835 114-8836 114-8837 114-8838 114-8839 114-8840 114-8841
ESD safety ankle boots, lace-up, L10, Vintage Pirate
Schoenen en laarzen
Vintage style ESD safety shoe made from waxy cowhide leather, with padded tongue and a dual-density VIBRAM® rubber/PU tread sole L10 PIRATE.
  • Fabric lining Bioactive
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO grey
  • Anatomically shaped insole
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Grey/red
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC