Microscope objective lens
Leverancier: LEICA MICROSYSTEMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11506236 11506241 11506271 11506272 11506369 11506273 11506408 11506298 11506203 11506402 11506321 11506276 11506237 11506386 11506406 11506248 11506217 11506097 11506382 11506519 11506525 11506302 11506158 11506405 11506096 11506349 11556036 11556060 11556061 11556065 11556071 11556511 11506083 11506184 11506220 11506227 11506401 11506277 11506526 11566069 11566070 11566071 11566072 11566073 11506243 11506314 11506385 11566077 11566083 11566067
LEIM11506236EA 245 EUR
LEIM11506236 LEIM11506241 630-3303 630-3304 630-3305 LEIM11506273 630-3302 630-3301 630-3309 630-3310 LEIM11506321 LEIM11506276 LEIM11506237 LEIM11506386 630-3189 630-3188 LEIM11506217 LEIM11506097 LEIM11506382 LEIM11506519 LEIM11506525 LEIM11506302 LEIM11506158 LEIM11506405 LEIM11506096 LEIM11506349 LEIM11556036 LEIM11556060 LEIM11556061 LEIM11556065 LEIM11556071 LEIM11556511 LEIM11506083 LEIM11506184 LEIM11506220 LEIM11506227 LEIM11506401 LEIM11506277 LEIM11506526 630-3366 630-3367 630-3368 630-3369 630-3370 LEIM11506243 LEIM11506314 LEIM11506385 LEIM11566077 LEIM11566083 LEIM11566067
Microscope objective lens
Microscoopobjectieven
The objective lens is a critical part of the microscope optics. The microscope objective is positioned near the sample, specimen, or object being observed. It has a very important role in imaging, as it forms the first magnified image of the sample.
  • The objective forms the first magnified image of the sample
The numerical aperture (NA) of the objective indicates its ability to gather light and largely determines the microscope’s resolution, the ability to distinguish fine details of the sample.
Objectives are categorised into performance classes onMore Product Information
Order Now
Specificaties
Environmentally Preferable