VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99102.230EA 292 EUR
99102.230 99103.230
VWR®, Cold Filter Plugging Point (CFPP) standards, Certified Reference Material
Standaarden Standaarden voor de petroleumindustrie
Cold Filter Plugging Point (CFPP) Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Cold Filter Plugging Point (CFPP) according to IP 309, EN 116 and ASTM D6371 also other equivalent test methodology.
  • Tested in accordance with ASTM / EN or IP test method protocols
  • Low level of uncertainty achieves maximum data accuracy
  • Supplied in tamper-evident UN compliant security pack with 12 month shelf life
Certificaten: Fully certified in accordance with ISO 17034 under accreditation by UKAS.