PETG certified clean biotainer bottles, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
383005-42 383025-42 383110-42 383230-20 383230-42 383415-16 383415-42
216-2272EA 1377 EUR
216-2272 216-2271 216-2273 216-2274 216-2275 216-2276 216-2277
PETG certified clean biotainer bottles, Nalgene®
Flessen Kunststof flessen
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Sterile
  • Clear finish
  • PETG construction
Nalgene™ certified biotainer bottles reduce the risk of contamination.
Certificaten: Validated to USP 788 and <0,5 EU/ml endotoxin.