Square PETG platinum certified clean biotainer bottles, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CE-N3005-42 CE-N3025-42 CE-N3110-42 CE-N3230-42
216-2313EA 2163 EUR
216-2313 216-2310 216-2314 216-2315
Square PETG platinum certified clean biotainer bottles, Nalgene®
Flessen Kunststof flessen
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Durable, translucent PETG construction ensures the integrity of samples
  • Closures feature a silicone liner for leakproof assurance
  • Customization options are available for fluid transfer applications
  • Extensive validation documentation support
  • Suitable for use in storage conditions of –100 to 100 °C