Square PETG platinum certified clean diagnostic bottles, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CE-N2035-0005 CE-N2035-0020
216-2296EA 14153 EUR
216-2296 216-2297
Square PETG platinum certified clean diagnostic bottles, Nalgene®
Flessen Kunststof flessen
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Non-pyrogenic and non-cytotoxic
  • 20-415 lined white HDPE closure attached
  • Leakproof
Bottles are washed with USP water for injection and handled post-washing in Class 5 cleanrooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than ¹/₃ the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level (SAL) of 10⁻⁶ inside the container/closure, including two heat-sealed sterile barriers to protect the product prior to use.
Certificaten: ISO 13485:2003, 21CFR177.1520, USP Class VI.
Verpakking: Diagnostic bottles are are packed in shrink-wrapped trays with closures attached.