PC certified clean validation bottles, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
383500-05 383500-20
216-2287EA 7480 EUR
216-2287 216-2288
PC certified clean validation bottles, Nalgene®
Flessen Kunststof flessen
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Polycarbonate construction
  • Sterile
  • Clear finish
  • Certified clean
These bottles can reduce the risk of contamination.
Certificaten: Validated to USP 788 and <0,5 EU/ml endotoxin.